Stampa…

Hexyl nicotinate ≥98.0% (by GC)

Fornitore: TCI
Attenzione

Synonyms: Nicotinic Acid Hexyl Ester

N0329-25G N0329-5G
TCIAN0329-25GEA 83 EUR
TCIAN0329-25G TCIAN0329-5G
Hexyl nicotinate ≥98.0% (by GC)
Hexyl nicotinate
Formula: C₁₂H₁₇NO₂
Punto di ebollizione: 104°C
MDL Number: MFCD00023587
CAS Number: 23597-82-2
EINECS: 245-767-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=219cea2b86152f836b1a6a5cd16596b0b349cd6b&vid=219cea2b86152f836b1a6a5cd16596b0b349cd6b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR