Stampa…

Basic Blue 24 (New Methylene Blue)

Fornitore: TCI
Attenzione

N0318-25G N0318-5G
TCIAN0318-25GEA 196 EUR
TCIAN0318-25G TCIAN0318-5G
Basic Blue 24 (New Methylene Blue)
Basic Blue 24 (New Methylene Blue)
Formula: C₁₈H₂₂ClN₃S·0.5 ZnCl₂
MDL Number: MFCD00135878
CAS Number: 6586-05-6
EINECS: 229-516-6

Visualizza i codici

Risultati del test di specificazione

Appearance Dark green to Dark red to Black powder to crystal
Lambda max. 626.0 to 630.0 nm (EtOH)
Absorbance min. 0.800 (4mg/L, EtOH, 626.0 to 630.0 nm) (calcd.on dry.substance)
Drying loss max. 5.0 %

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR