Stampa…

Nile blue chloride (Basic blue 12)

Fornitore: TCI EUROPE

Synonyms: Nile Blue A chloride, 5-Amino-9-(diethylamino)benzo[a]phenoxazin-7-ium chloride

N0317-25G N0317-5G
TCIAN0317-25GEA 116 EUR
TCIAN0317-25G TCIAN0317-5G
Nile blue chloride (Basic blue 12)
Nile blue chloride (Basic blue 12)
Formula: C₂₀H₂₀ClN₃O
Peso molecolare: 353,85 g/mol
MDL Number: MFCD00011936
CAS Number: 2381-85-3
EINECS: 219-181-4

Visualizza i codici

Risultati del test di specificazione

Appearance Green to Dark green powder to crystal
Lambda max. 632.0 to 638.0 nm (EtOH (50%))
Absorbance (E1%1cm) min. 1500 (EtOH (50%), 632.0 to 638.0 nm)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR