1-Nonanesulphonic acid sodium salt ≥98.0% (by titrimetric analysis)
Fornitore: TCI
Formula:
C₉H₁₉NaO₃S |
MDL Number:
MFCD00025010 CAS Number: 35192-74-6 |
|
Risultati del test di specificazione
Appearance | White powder to crystal |
Purity (Ion exchange titration) | min. 98.0 % |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...