Stampa…

4-Nitro-1-naphthylamine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Nitronaphthalen-1-amine, 1-Amino-4-nitronaphthalene

N0213-25G N0213-5G
TCIAN0213-25GEA 168 EUR
TCIAN0213-25G TCIAN0213-5G
4-Nitro-1-naphthylamine ≥98.0% (by GC)
4-Nitro-1-naphthylamine
Formula: C₁₀H₈N₂O₂
Peso molecolare: 188,19 g/mol
MDL Number: MFCD00004026
CAS Number: 776-34-1
EINECS: 212-277-7

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 193.0 to 196.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR