Stampa…

1-Naphthylhydrazine hydrochloride ≥90.0% (by HPLC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Naphthalen-1-ylhydrazine hydrochloride

N0064-1G N0064-5G
TCIAN0064-1GEA 27 EUR
TCIAN0064-1G TCIAN0064-5G
1-Naphthylhydrazine hydrochloride ≥90.0% (by HPLC)
1-Naphthylhydrazine hydrochloride
Formula: C₁₀H₁₀N₂
Peso molecolare: 194,66 g/mol
MDL Number: MFCD00060157
CAS Number: 2243-56-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light gray to Light orange powder to crystal
Purity (HPLC) min. 90.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR