Stampa…

Methyl 4-Isopropylbenzoate ≥98.0% (by GC)

Fornitore: TCI

M3307-1G M3307-5G
TCIAM3307-1GEA 67 EUR
TCIAM3307-1G TCIAM3307-5G
Methyl 4-Isopropylbenzoate ≥98.0% (by GC)
Methyl 4-Isopropylbenzoate
Formula: C₁₁H₁₄O₂
Punto di ebollizione: 126°C
MDL Number: MFCD06203666
CAS Number: 20185-55-1

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR