Stampa…

Moguisteine ≥98.0% (by HPLC, total nitrogen)

Fornitore: TCI

M3305-1G M3305-250MG
TCIAM3305-1GEA 201 EUR
TCIAM3305-1G TCIAM3305-250MG
Moguisteine ≥98.0% (by HPLC, total nitrogen)
Moguisteine
Formula: C₁₆H₂₁NO₅S
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00866839
CAS Number: 119637-67-1

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 58.0 to 62.0 deg-C
Specific rotation [a]20/D -1.0 to +1.0 deg (C=1, CHCL3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR