Stampa…

Mitotane ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: o,p'-DDD, 2,4'-TDE, o,p'-TDE, 2,4'-DDD

M3304-1G M3304-5G
TCIAM3304-1GEA 46 EUR
TCIAM3304-1G TCIAM3304-5G
Mitotane ≥98.0% (by GC)
Mitotane
Formula: C₁₄H₁₀Cl₄
Peso molecolare: 320,04 g/mol
MDL Number: MFCD00000850
CAS Number: 53-19-0
Merck Index: 14,06216

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 76.0 to 80.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR