Stampa…

3-Mercapto-2-methyl-1-pentanol mixture of diasteroisomers ≥97.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Methyl-3-sulphanyl-1-pentanol

M3274-1G M3274-5G
TCIAM3274-1GEA 29 EUR
TCIAM3274-1G TCIAM3274-5G
3-Mercapto-2-methyl-1-pentanol mixture of diasteroisomers ≥97.0% (by GC, titration analysis)
3-Mercapto-2-methyl-1-pentanol
Formula: C₆H₁₄OS
Peso molecolare: 134,24 g/mol
Punto di ebollizione: 46°C
MDL Number: MFCD08741458
CAS Number: 227456-27-1
EINECS: 434-900-3

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 % (total of isomer)
Purity (Iodometric Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR