4'-Methyl-4-biphenylboronic acid (contains varying amounts of Anhydride)
Fornitore: TCI
|
Attenzione
|
M3235-1G
M3235-5G
TCIAM3235-1GEA
53
EUR
TCIAM3235-1G
TCIAM3235-5G
4'-Methyl-4-biphenylboronic acid (contains varying amounts of Anhydride)
4'-Methyl-4-biphenylboronic acid
Formula:
C₁₃H₁₃BO₂ Punto di fusione: 138°C |
MDL Number:
MFCD04038170 CAS Number: 393870-04-7 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 110.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...