2-(Methoxycarbonyl)phenylboronic acid pinacol ester ≥98.0% (by GC)
Fornitore: TCI
Synonyms:
2-(2-Methoxycarbonylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid methyll ester, 2-(Methoxycarbonyl)phenylboronic acid Pinacol ester, Methyl 2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 2-Methoxycarbonylphenylboronic acid pinacol ester
Formula:
C₁₄H₁₉BO₄ |
MDL Number:
MFCD05663866 CAS Number: 653589-95-8 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 43.0 to 47.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...