Stampa…
Methyl-4-bromo-2-fluorobenzoate ≥98.0% (by GC)
Fornitore: TCI
Attenzione
Synonyms: Methyl 4-bromo-2-fluorobenzoate
M3229-5G M3229-25G
TCIAM3229-5GEA 46 EUR
TCIAM3229-5G TCIAM3229-25G
Methyl-4-bromo-2-fluorobenzoate ≥98.0% (by GC)
Methyl-4-bromo-2-fluorobenzoate
Formula: C₈H₆BrFO₂
Peso molecolare: 233.04 g/mol
MDL Number: MFCD06203731
CAS Number: 179232-29-2
Visualizza i codici
Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 58.0 to 62.0 deg-C