Stampa…

1-Methylimidazole bis(trifluoromethanesulphonyl)imide ≥97.0% (by HPLC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1-Methylimidazole Bis(trifluoromethanesulphonyl)imide, 1-Methylimidazolium bis(trifluoromethanesulphonyl)imide

M3210-5G M3210-25G
TCIAM3210-5GEA 34 EUR
TCIAM3210-5G TCIAM3210-25G
1-Methylimidazole bis(trifluoromethanesulphonyl)imide ≥97.0% (by HPLC)
1-Methylimidazole bis(trifluoromethanesulphonyl)imide
Formula: C₆H₇F₆N₃O₄S₂
Peso molecolare: 363,25 g/mol
MDL Number: MFCD18251286
CAS Number: 353239-08-4

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Melting point 47.0 to 51.0 deg-C
Water max. 1.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR