Stampa…

Methyl 4-Formyl-3-nitrobenzoate ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: 4-Formyl-3-nitrobenzoic acid methyl ester

M3198-1G M3198-5G
TCIAM3198-1GEA 31 EUR
TCIAM3198-1G TCIAM3198-5G
Methyl 4-Formyl-3-nitrobenzoate ≥98.0% (by GC)
Methyl 4-Formyl-3-nitrobenzoate
Formula: C₉H₇NO₅
Peso molecolare: 209,16 g/mol
MDL Number: MFCD04039968
CAS Number: 153813-69-5

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 62.0 to 66.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR