2-Methyl-3-butenoic acid ≥96.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
2-Methylbut-3-enoic acid
M3188-1G
M3188-5G
TCIAM3188-1GEA
69
EUR
TCIAM3188-1G
TCIAM3188-5G
2-Methyl-3-butenoic acid ≥96.0% (by GC, titration analysis)
2-Methyl-3-butenoic acid
Formula:
C₅H₈O₂ Peso molecolare: 100,12 g/mol Punto di ebollizione: 94°C |
MDL Number:
MFCD00042650 CAS Number: 53774-20-2 UN: 3265 ADR: 8,II |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 96.0 % |
Purity (Neutralization titration) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...