Methyl-3-(benzylamino)propanoate 98,0 GC, T
Fornitore: TCI
Synonyms:
Methyl 3-(benzylamino)propanoate
Formula:
C₁₁H₁₅NO₂ Peso molecolare: 193.25 g/mol Punto di ebollizione: 147°C |
MDL Number:
MFCD01475856 CAS Number: 23574-01-8 |
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...