Stampa…

(±)-Methyl-2,3-dibromopropionate ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2,3-Dibromopropionic acid methyll ester, Methyl 2,3-Dibromopropionate, Methyl 2,3-dibromopropanoate

M3182-25G M3182-5G
TCIAM3182-25GEA 85 EUR
TCIAM3182-25G TCIAM3182-5G
(±)-Methyl-2,3-dibromopropionate ≥98.0% (by GC)
(±)-Methyl-2,3-dibromopropionate
Formula: C₄H₆Br₂O₂
Peso molecolare: 245,9 g/mol
Punto di ebollizione: 204°C
MDL Number: MFCD00017882
CAS Number: 1729-67-5
EINECS: 217-044-3

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR