Mitiglinide calcium
Fornitore: TCI
Synonyms:
Calcium (S)-2-benzyl-4-((3aR,7aS)-octahydro-2H-isoindol-2-yl)-4-oxobutanoate, S21403, (2S)-4-[(3aR,7aS)-1,3,3a,4,5,6,7,7a-octahydroisoindol-2-yl]-2-benzyl-4-oxobutanoate, calcium
Formula:
C₃₈H₄₈CaN₂O₆ Peso molecolare: 668.87 g/mol Punto di fusione: 185°C |
MDL Number:
MFCD09955408 CAS Number: 145525-41-3 Merck Index: 14,06211 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Chelometric Titration) | min. 97.0 % |
Specific rotation [a]20/D | +5.0 to +9.0 deg (C=1, MeOH (calcd.on anh.substance)) |
Water | 4.5 to 6.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...