Stampa…

5-(4-Methylpiperazin-1-yl)pyridin-2-amine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

Synonyms: 1-Methyl-4-(6-aminopyridin-3-yl)piperazine

M3176-1G M3176-5G
TCIAM3176-1GEA 54 EUR
TCIAM3176-1G TCIAM3176-5G
5-(4-Methylpiperazin-1-yl)pyridin-2-amine ≥98.0% (by GC)
5-(4-Methylpiperazin-1-yl)pyridin-2-amine
Formula: C₁₀H₁₆N₄
Peso molecolare: 192,2608 g/mol
MDL Number: MFCD10009563
CAS Number: 571189-49-6
UN: 3263
ADR: 8,II

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 146.0 to 150.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR