SMER-3 ≥97.0% (by HPLC)
Fornitore: TCI EUROPE
Synonyms:
9H-Indeno[1,2-b][1,2,5]oxadiazolo[3,4-e]pyrazin-9-one
M3172-10MG
M3172-50MG
TCIAM3172-10MGEA
102
EUR
TCIAM3172-10MG
TCIAM3172-50MG
SMER-3 ≥97.0% (by HPLC)
SMER-3
Formula:
C₁₁H₄N₄O₂ Peso molecolare: 224,18 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00451458 CAS Number: 67200-34-4 |
Risultati del test di specificazione
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Elemental analysis (Nitrogen) | 23.70 to 26.00 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...