1-Methyl-4-nitroimidazole ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
M3171-1G
M3171-5G
TCIAM3171-1GEA
71
EUR
TCIAM3171-1G
TCIAM3171-5G
1-Methyl-4-nitroimidazole ≥98.0% (by GC)
1-Methyl-4-nitroimidazole
Formula:
C₄H₅N₃O₂ Peso molecolare: 127,10 g/mol |
MDL Number:
MFCD00159683 CAS Number: 3034-41-1 |
Risultati del test di specificazione
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 134.0 to 138.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...