Stampa…

Methyl-2-naphthoate ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Methyl 2-naphthoate

M3169-5G M3169-25G
TCIAM3169-5GEA 130 EUR
TCIAM3169-5G TCIAM3169-25G
Methyl-2-naphthoate ≥98.0% (by GC)
Methyl-2-naphthoate
Formula: C₁₂H₁₀O₂
Peso molecolare: 186,21 g/mol
Punto di ebollizione: 290°C
MDL Number: MFCD00017345
CAS Number: 2459-25-8

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 76.0 to 80.0 deg-C
Solubility in Chloroform almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR