6-Methoxytryptamine ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
M3162-100MG
M3162-500MG
TCIAM3162-100MGEA
89
EUR
TCIAM3162-100MG
TCIAM3162-500MG
6-Methoxytryptamine ≥98.0% (by HPLC, titration analysis)
6-Methoxytryptamine
Formula:
C₁₁H₁₄N₂O Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00005663 CAS Number: 3610-36-4 EINECS: 222-778-2 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 141.0 to 145.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...