1-(2-Methoxyphenyl)-2-thiourea ≥98.0% (by HPLC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
N-(2-Methoxyphenyl)thiourea, 1-(2-Methoxyphenyl)thiourea, 2-Methoxyphenylthiourea
M3158-5G
M3158-25G
TCIAM3158-5GEA
42
EUR
TCIAM3158-5G
TCIAM3158-25G
1-(2-Methoxyphenyl)-2-thiourea ≥98.0% (by HPLC)
1-(2-Methoxyphenyl)-2-thiourea
Formula:
(C₆H₄(OCH₃))NC(S)NH₂ Peso molecolare: 182,25 g/mol |
MDL Number:
MFCD00022162 CAS Number: 1516-37-6 UN: 2811 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 148.0 to 152.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...