Stampa…

Methyl-4-aminobutanoate hydrochloride ≥98.0% (by total nitrogen and titration analysis)

Fornitore: TCI EUROPE

Synonyms: H-γ-Abu-OMe.HCl

M3154-25G M3154-5G
TCIAM3154-25GEA 154 EUR
TCIAM3154-25G TCIAM3154-5G
Methyl-4-aminobutanoate hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Methyl-4-aminobutanoate hydrochloride
Formula: C₅H₁₁NO₂·HCl
Peso molecolare: 153,61 g/mol
MDL Number: MFCD00043270
CAS Number: 13031-60-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Purity (with Total Nitrogen) min. 98.0 %
Melting point 122.0 to 126.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR