1-Methoxy-4-(trans-4-propylcyclohexyl)benzene ≥98.0% (by GC)
Fornitore: TCI
Formula:
C₁₆H₂₄O |
MDL Number:
MFCD06658193 CAS Number: 81936-32-5 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to lump |
Purity (GC) | min. 98.0 % |
Melting point | 31.0 to 35.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...