Stampa…

Methyl-2-bromo-4-chlorobenzoate ≥98.0% (by GC)

Fornitore: TCI
Attenzione

M3131-25G M3131-5G
TCIAM3131-25GEA 185 EUR
TCIAM3131-25G TCIAM3131-5G
Methyl-2-bromo-4-chlorobenzoate ≥98.0% (by GC)
Methyl-2-bromo-4-chlorobenzoate
Formula: C₈H₆BrClO₂
MDL Number: MFCD10566822
CAS Number: 57381-62-1

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR