4-(Morpholinomethyl)phenylboronic acid (contains varying amounts of Anhydride)
Fornitore: TCI
|
Attenzione
|
Formula:
C₁₁H₁₆BNO₃ Punto di fusione: 85°C |
MDL Number:
MFCD06801707 CAS Number: 279262-23-6 |
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Neutralization titration) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...