Stampa…

Methyl-2-amino-4-bromobenzoate ≥97.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Amino-4-bromobenzoic acid methyll ester, Methyl 2-amino-4-bromobenzoate

M3125-5G M3125-25G
TCIAM3125-5GEA 58 EUR
TCIAM3125-5G TCIAM3125-25G
Methyl-2-amino-4-bromobenzoate ≥97.0% (by GC)
Methyl-2-amino-4-bromobenzoate
Formula: C₈H₈BrNO₂
Peso molecolare: 230,0586 g/mol
MDL Number: MFCD07783014
CAS Number: 135484-83-2

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 77.0 to 81.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR