Stampa…

2-[2-(2-Methoxyethoxy)ethoxy]ethyl 4-Methylbenzenesulphonate ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: 2-[2-(2-Methoxyethoxy)ethoxy]ethyl p-Toluenesulphonate, 2-[2-(2-Methoxyethoxy)ethoxy]ethyl Tosylate, 4-Methylbenzenesulphonic acid 2-[2-(2-Methoxyethoxy)ethoxy]ethyl ester, m-PEG3-Tos

M3122-1G M3122-5G
TCIAM3122-1GEA 78 EUR
TCIAM3122-1G TCIAM3122-5G
2-[2-(2-Methoxyethoxy)ethoxy]ethyl 4-Methylbenzenesulphonate ≥98.0% (by HPLC, titration analysis)
2-[2-(2-Methoxyethoxy)ethoxy]ethyl 4-Methylbenzenesulphonate
Formula: C₁₄H₂₂O₆S
Peso molecolare: 318,38 g/mol
CAS Number: 62921-74-8

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (HPLC) min. 98.0 area%
Purity (Ester Value) min. 98.0 %
Acid Value max. 3.0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR