MHY1485 ≥95.0% (by HPLC)
Fornitore: TCI EUROPE
Synonyms:
4,6-Dimorpholino-N-(4-nitrophenyl)-1,3,5-triazin-2-amine
M3107-100MG
M3107-25MG
TCIAM3107-100MGEA
209
EUR
TCIAM3107-100MG
TCIAM3107-25MG
MHY1485 ≥95.0% (by HPLC)
MHY1485
Formula:
C₁₇H₂₁N₇O₄ Peso molecolare: 387,39 g/mol Punto di fusione: 259°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00489974 CAS Number: 326914-06-1 |
Risultati del test di specificazione
Appearance | White to Amber powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Elemental analysis (Nitrogen) | 24.00 to 26.50 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...