Stampa…

4-Methyl-2-(p-tolyl)pyridine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Methyl-2-(4-methylphenyl)pyridine

M3105-200MG M3105-1G
TCIAM3105-200MGEA 32 EUR
TCIAM3105-200MG TCIAM3105-1G
4-Methyl-2-(p-tolyl)pyridine ≥98.0% (by GC, titration analysis)
4-Methyl-2-(p-tolyl)pyridine
Formula: C₁₃H₁₃N
Peso molecolare: 183,25 g/mol
MDL Number: MFCD09031945
CAS Number: 80635-92-3

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 50.0 to 54.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR