Methyl-2-chloro-4-fluorobenzoate ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Methyl 2-chloro-4-fluorobenzenecarboxylate, Methyl 2-chloro-4-fluorobenzoate, 2-Chloro-4-fluorobenzoic acid methyl ester
M3102-5G
M3102-25G
TCIAM3102-5GEA
29
EUR
TCIAM3102-5G
TCIAM3102-25G
Methyl-2-chloro-4-fluorobenzoate ≥98.0% (by GC)
Methyl-2-chloro-4-fluorobenzoate
Formula:
C₈H₆ClFO₂ Peso molecolare: 188,59 g/mol |
MDL Number:
MFCD00173937 CAS Number: 85953-29-3 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Acid Value | max. 2.0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...