4-Morpholinobenzoic acid ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
4-Morpholinobenzoic acid, 4-(4-Morpholinyl)benzoic acid
M3100-1G
M3100-5G
TCIAM3100-1GEA
26
EUR
TCIAM3100-1G
TCIAM3100-5G
4-Morpholinobenzoic acid ≥98.0% (by GC, titration analysis)
4-Morpholinobenzoic acid
Formula:
C₁₁H₁₃NO₃ Peso molecolare: 207,2300 g/mol Punto di fusione: 277°C |
MDL Number:
MFCD00588379 CAS Number: 7470-38-4 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...