Stampa…

N-Methoxy-N,4-dimethylbenzamide ≥95.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

M3098-1G M3098-5G
TCIAM3098-1GEA 75 EUR
TCIAM3098-1G TCIAM3098-5G
N-Methoxy-N,4-dimethylbenzamide ≥95.0% (by GC)
N-Methoxy-N,4-dimethylbenzamide
Formula: C₁₀H₁₃NO₂
Peso molecolare: 179,22 g/mol
MDL Number: MFCD02684311
CAS Number: 122334-36-5

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR