Stampa…

N-Methoxy-N-methylbutyramide ≥98.0% (by GC)

Fornitore: TCI
Attenzione

Synonyms: N-Methoxy-N-methylbutanamide

M3097-1G M3097-5G
TCIAM3097-1GEA 53 EUR
TCIAM3097-1G TCIAM3097-5G
N-Methoxy-N-methylbutyramide ≥98.0% (by GC)
N-Methoxy-N-methylbutyramide
Formula: C₆H₁₃NO₂
MDL Number: MFCD14707556
CAS Number: 109480-78-6
UN: 1993
ADR: 3,III

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR