Sophoridine ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
(7aR,13aR,13bR,13cS)-Dodecahydro-1H,5H,10H-dipyrido[2,1-f:3',2',1'-ij][1,6]naphthyridin-10-one, (5beta)-Matridin-15-one, (41S,7aR,13aR,13bR)-Dodecahydro-1H-dipyrido[2,1-f:3',2',1'-ij][1,6]naphthyridin-10(41H)-one
M3093-250MG
M3093-50MG
TCIAM3093-250MGEA
197
EUR
TCIAM3093-250MG
TCIAM3093-50MG
Sophoridine ≥98.0% (by GC, titration analysis)
Sophoridine
Formula:
C₁₅H₂₄N₂O Peso molecolare: 248,37 g/mol Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00238686 CAS Number: 6882-68-4 UN: 1544 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 107.0 to 111.0 deg-C |
Specific rotation [a]20/D | -66.0 to -73.0 deg (C=1, ethanol) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...