Stampa…

Methyl-5-bromo-2-iodobenzoate ≥95.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: Methyl 5-bromo-2-iodobenzoate

M3084-1G M3084-5G
TCIAM3084-1GEA 16 EUR
TCIAM3084-1G TCIAM3084-5G
Methyl-5-bromo-2-iodobenzoate ≥95.0% (by GC)
Methyl-5-bromo-2-iodobenzoate
Formula: C₈H₆BrIO₂
Peso molecolare: 340,9400 g/mol
Punto di ebollizione: 134°C
MDL Number: MFCD00144771
CAS Number: 181765-86-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=face72934bed3422b3e9a9f660c706ecfc84eae8&vid=face72934bed3422b3e9a9f660c706ecfc84eae8&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 95.0 %
Melting point 43.0 to 47.0 deg-C
Acid Value max. 3.0

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR