Meisoindigo ≥98.0% (by HPLC)
Fornitore: TCI
Formula:
C₁₇H₁₂N₂O₂ Punto di fusione: 230°C |
MDL Number:
MFCD05988503 CAS Number: 97207-47-1 |
|
Risultati del test di specificazione
Appearance | Orange to Brown to Dark red powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Nitrogen) | 9.50 to 10.50 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...