Mal-amido-PEG2-NHS ≥98.0% (by GC, total nitrogen)
Fornitore: TCI
Formula:
C₁₈H₂₃N₃O₉ Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD11041137 CAS Number: 955094-26-5 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 90.0 to 94.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...