Stampa…

1-Methyl-3-propylimidazolium bis(trifluoromethylsulphonyl)imide ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1-Methyl-3-propylimidazolium Bis(trifluoromethanesulphonyl)imide

M3059-5G M3059-25G
TCIAM3059-5GEA 34 EUR
TCIAM3059-5G TCIAM3059-25G
1-Methyl-3-propylimidazolium bis(trifluoromethylsulphonyl)imide ≥98.0% (by HPLC, titration analysis)
1-Methyl-3-propylimidazolium bis(trifluoromethylsulphonyl)imide
Formula: C₉H₁₃F₆N₃O₄S₂
Peso molecolare: 405,34 g/mol
MDL Number: MFCD15071469
CAS Number: 216299-72-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e5b2ce849069240868e3e97102b599e25008926e&vid=e5b2ce849069240868e3e97102b599e25008926e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (HPLC) min. 98.0 area%
Purity (Ion exchange titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR