4-Methylthiophene-2-carboxaldehyde ≥95.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
2-Formyl-4-methylthiophene, 4-Methylthiophene-2-carbaldehyde
M3054-1G
M3054-5G
TCIAM3054-1GEA
54
EUR
TCIAM3054-1G
TCIAM3054-5G
4-Methylthiophene-2-carboxaldehyde ≥95.0% (by GC)
4-Methylthiophene-2-carboxaldehyde
Formula:
C₆H₆OS Peso molecolare: 126,17 g/mol Punto di ebollizione: 86°C Flash Pt: 92°C |
MDL Number:
MFCD00159561 CAS Number: 6030-36-0 |
Risultati del test di specificazione
Appearance | Colorless to Red to Green clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...