Stampa…

(S)-(-)-N,α-Dimethylbenzylamine ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Pericolo

Synonyms: (S)-N,α-Dimethylbenzylamine, (S)-(-)-alpha-Methyl-N-methylbenzylamine, (S)-(-)-N-Methyl-1-phenylethylamine, (S)-(-)-N α-Dimethylbenzylamine

M3047-1ML M3047-5ML
TCIAM3047-1MLEA 50 EUR
TCIAM3047-1ML TCIAM3047-5ML
(S)-(-)-N,α-Dimethylbenzylamine ≥98.0% (by GC, titration analysis)
(S)-(-)-N,α-Dimethylbenzylamine
Formula: C₉H₁₃N
Peso molecolare: 135,2100 g/mol
Punto di ebollizione: 79°C
Flash Pt: 61°C
MDL Number: MFCD00067113
CAS Number: 19131-99-8
UN: 2735
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f753d3ce631746570f9d96a482730a287c164ce&vid=3f753d3ce631746570f9d96a482730a287c164ce&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -73.0 to -77.0 deg (C=2,CHCL3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR