Stampa…

N-Methyl-3-nitroaniline ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

M3042-1G M3042-5G
TCIAM3042-1GEA 130 EUR
TCIAM3042-1G TCIAM3042-5G
N-Methyl-3-nitroaniline ≥98.0% (by GC, titration analysis)
N-Methyl-3-nitroaniline
Formula: C₇H₈N₂O₂
Peso molecolare: 152,153 g/mol
MDL Number: MFCD00963641
CAS Number: 619-26-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ccabcb6825ac09270732a7f47f982fedbd74ffb&vid=2ccabcb6825ac09270732a7f47f982fedbd74ffb&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 65.0 to 69.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR