Stampa…

1-Methyl-3-propylimidazolium chloride ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1-Methyl-3-propylimidazolium Chloride

M3035-5G M3035-25G
TCIAM3035-5GEA 35 EUR
TCIAM3035-5G TCIAM3035-25G
1-Methyl-3-propylimidazolium chloride ≥98.0% (by HPLC, titration analysis)
1-Methyl-3-propylimidazolium chloride
Formula: C₇H₁₃ClN₂
Peso molecolare: 160,65 g/mol
MDL Number: MFCD17014728
CAS Number: 79917-89-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1b8397624d2d31eea4e78802314d7d47e63097e&vid=b1b8397624d2d31eea4e78802314d7d47e63097e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR