Stampa…

1-Methyl-3-propylimidazolium bromide ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI
Attenzione

Synonyms: 1-propenyl-3-methylimidazolium bromide

M3034-5G M3034-25G
TCIAM3034-5GEA 46 EUR
TCIAM3034-5G TCIAM3034-25G
1-Methyl-3-propylimidazolium bromide ≥98.0% (by HPLC, titration analysis)
1-Methyl-3-propylimidazolium bromide
Formula: C₇H₁₃BrN₂
Peso molecolare: 205.10 g/mol
MDL Number: MFCD17014727
CAS Number: 85100-76-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73fd5e9112202dda4612e3846bb38d0f81996158&vid=73fd5e9112202dda4612e3846bb38d0f81996158&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Orange clear liquid
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR