tert-Butyl hydrogen succinate ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Succinic acid mono-tert-butyl ester, 4-(tert-Butoxy)-4-oxobutanoic acid, tert-Butyl Hydrogen Succinate, Mono-tert-butyl Succinate
M3033-1G
M3033-5G
TCIAM3033-1GEA
24
EUR
TCIAM3033-1G
TCIAM3033-5G
tert-Butyl hydrogen succinate ≥98.0% (by GC, titration analysis)
tert-Butyl hydrogen succinate
Formula:
C₈H₁₄O₄ Peso molecolare: 174,196 g/mol Punto di ebollizione: 92°C |
MDL Number:
MFCD00273441 CAS Number: 15026-17-2 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 51.0 to 55.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...