Stampa…

4-Methylumbelliferyl-Β-D-cellobiopyranoside ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: 4-Methylumbelliferyl beta-D-Cellobioside

M3028-100MG M3028-500MG
TCIAM3028-100MGEA 153 EUR
TCIAM3028-100MG TCIAM3028-500MG
4-Methylumbelliferyl-Β-D-cellobiopyranoside ≥98.0% (by HPLC)
4-Methylumbelliferyl-Β-D-cellobiopyranoside
Formula: C₂₂H₂₈O₁₃
Peso molecolare: 500,45 g/mol
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00063279
CAS Number: 72626-61-0
EINECS: 276-744-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dde3ec816047dfdf231976861a8513642a4aada9&vid=dde3ec816047dfdf231976861a8513642a4aada9&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Specific rotation [a]20/D -92.0 to -97.0 deg (C=0.2, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR