Stampa…

4-Methylumbelliferyl-Α-L-arabinopyranoside ≥98.0% (by HPLC)

Fornitore: TCI EUROPE

Synonyms: 4-Methylumbelliferyl alpha-L-Arabinopyranoside

M3025-50MG M3025-250MG
TCIAM3025-50MGEA 69 EUR
TCIAM3025-50MG TCIAM3025-250MG
4-Methylumbelliferyl-Α-L-arabinopyranoside ≥98.0% (by HPLC)
4-Methylumbelliferyl-Α-L-arabinopyranoside
Formula: C₁₅H₁₆O₇
Peso molecolare: 308,28 g/mol
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00064087
CAS Number: 69414-26-2
EINECS: 273-991-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=190e3a759a5187780e68e522b5fd626b2a1d5864&vid=190e3a759a5187780e68e522b5fd626b2a1d5864&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Carbon) 57.50 to 59.50 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR